Skip to Content

BDPH9BCDC5AE

Bide Pharmatech

2-Bromo-5-(difluoromethyl)-1,3,4-thiadiazole

Synonym(s):

2-bromo-5-(difluoromethyl)-1,3,4-thiadiazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C3H1N2Br1S1F2
CAS Number:
Molecular Weight:
215.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C3HBrF2N2S/c4-3-8-7-2(9-3)1(5)6/h1H

Inchi Key

GQUADXWTZXHNAT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-Solid or Liquid or Lump



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library