Skip to Content

BDPH9BCDC65F

Bide Pharmatech

1-(3,5-Dichlorophenyl)thiourea

Synonym(s):

N-(3,5-dichlorophenyl)thiourea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H6N2Cl2S1
CAS Number:
Molecular Weight:
221.11
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H6Cl2N2S/c8-4-1-5(9)3-6(2-4)11-7(10)12/h1-3H,(H3,10,11,12)

Inchi Key

BNJARXOEVLYKDN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 1-(3,5-Dichlorophenyl)thiourea


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Skull and crossbones


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library