Skip to Content
Merck

BDPH9BCDC7D2

Bide Pharmatech

4-(tert-Butoxy)-3-chloroaniline

Synonym(s):

4-tert-butoxy-3-chloroaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H14O1N1Cl1
CAS Number:
Molecular Weight:
199.68
MDL number:
Product BDPH9BCDC7D2 is not currently sold in your country.


Inchi Code

1S/C10H14ClNO/c1-10(2,3)13-9-5-4-7(12)6-8(9)11/h4-6H,12H2,1-3H3

Inchi Key

UUXBZGOILICMAR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 4-(tert-Butoxy)-3-chloroaniline



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library