Skip to Content

BDPH9BCDCBB1

Bide Pharmatech

(4-Bromothiophen-2-yl)boronic acid

Synonym(s):

4-bromo-2-thienylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H4O2Br1S1B1
CAS Number:
Molecular Weight:
206.86
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H4BBrO2S/c6-3-1-4(5(7)8)9-2-3/h1-2,7-8H

Inchi Key

JNBQQYFWHGVOBY-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under (4-Bromothiophen-2-yl)boronic acid


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library