Skip to Content
Merck

BDPH9BCDCBEB

Bide Pharmatech

2-Decyl-1H-imidazole

Synonym(s):

2-decyl-1H-imidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9BCDCBEB is not currently sold in your country.


Inchi Code

1S/C13H24N2/c1-2-3-4-5-6-7-8-9-10-13-14-11-12-15-13/h11-12H,2-10H2,1H3,(H,14,15)

Inchi Key

LYNVLWLRSACENL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-Decyl-1H-imidazole




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library