Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C9H7Cl/c1-2-8-3-5-9(7-10)6-4-8/h1,3-6H,7H2
Inchi Key
MBIGNFNSALGEMI-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid or Semi-solid or liquid or lump
Still not finding the right product?
Explore all of our products under 1-(Chloromethyl)-4-ethynylbenzene
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
