Skip to Content
Merck

BDPH9BCDCFB2

Bide Pharmatech

1-tert-Butyl 15-(2,5-dioxopyrrolidin-1-yl) pentadecanedioate

Synonym(s):

1-(tert-butyl) 15-(2,5-dioxopyrrolidin-1-yl) pentadecanedioate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C23H39O6N1
CAS Number:
Molecular Weight:
425.57
MDL number:
Product BDPH9BCDCFB2 is not currently sold in your country.


Inchi Code

1S/C23H39NO6/c1-23(2,3)29-21(27)15-13-11-9-7-5-4-6-8-10-12-14-16-22(28)30-24-19(25)17-18-20(24)26/h4-18H2,1-3H3

Inchi Key

MKNIKVLVLSJHJK-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library