Skip to Content
Merck

BDPH9BCDD0F7

Bide Pharmatech

2-Bromo-5-(cyclopropylmethoxy)pyrazine

Synonym(s):

2-bromo-5-(cyclopropylmethoxy)pyrazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O1N2Br1
CAS Number:
Molecular Weight:
229.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9BrN2O/c9-7-3-11-8(4-10-7)12-5-6-1-2-6/h3-4,6H,1-2,5H2

Inchi Key

DRQQPNNWAXZONT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library