Skip to Content
Merck

BDPH9BCDD7F6

Bide Pharmatech

Ethyl 2-(piperidin-3-yl)acetate hydrochloride

Synonym(s):

ethyl 3-piperidinylacetate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H18O2N1Cl1
Molecular Weight:
207.7
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H17NO2.ClH/c1-2-12-9(11)6-8-4-3-5-10-7-8;/h8,10H,2-7H2,1H3;1H

Inchi Key

NUFQPPPCFKZWRT-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library