Skip to Content
Merck

BDPH9BCDDBB3

Bide Pharmatech

Ethyl 7-methoxy-1H-indole-2-carboxylate

Synonym(s):

ethyl 7-methoxy-1H-indole-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H13O3N1
CAS Number:
Molecular Weight:
219.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H13NO3/c1-3-16-12(14)9-7-8-5-4-6-10(15-2)11(8)13-9/h4-7,13H,3H2,1-2H3

Inchi Key

BRDVNJZQPSSLMK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library