Skip to Content

BDPH9BCDDEC5

Bide Pharmatech

2-(4-Bromophenyl)-1,3,4-thiadiazole

Synonym(s):

2-(4-bromophenyl)-1,3,4-thiadiazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5N2Br1S1
CAS Number:
Molecular Weight:
241.11
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H5BrN2S/c9-7-3-1-6(2-4-7)8-11-10-5-12-8/h1-5H

Inchi Key

ONJVHBYVYOIMLD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library