Skip to Content
Merck

BDPH9BCDE4EE

Bide Pharmatech

tert-Butyl (2-(2-acetylhydrazinyl)-2-oxoethyl)carbamate

Synonym(s):

tert-butyl (2-(2-acetylhydrazinyl)-2-oxoethyl)carbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H17O4N3
CAS Number:
Molecular Weight:
231.25
MDL number:
Product BDPH9BCDE4EE is not currently sold in your country.


Inchi Code

1S/C9H17N3O4/c1-6(13)11-12-7(14)5-10-8(15)16-9(2,3)4/h5H2,1-4H3,(H,10,15)(H,11,13)(H,12,14)

Inchi Key

IJQRYEPGZZLZEY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library