Skip to Content
Merck

BDPH9BCDE768

Bide Pharmatech

8-Boc-2-oxo-8-azabicyclo[3.2.1]octane

Synonym(s):

tert-butyl (1S,5R)-2-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H19O3N1
CAS Number:
Molecular Weight:
225.29
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H19NO3/c1-12(2,3)16-11(15)13-8-4-6-9(13)10(14)7-5-8/h8-9H,4-7H2,1-3H3

Inchi Key

SJGXXUBCKFNNGB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library