Skip to Content
Merck

BDPH9BCDEEEF

Bide Pharmatech

1-((1-(3-Fluorophenyl)ethyl)amino)propan-2-ol

Synonym(s):

1-{[1-(3-fluorophenyl)ethyl]amino}-2-propanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H16O1N1F1
CAS Number:
Molecular Weight:
197.25
MDL number:
Product BDPH9BCDEEEF is not currently sold in your country.


Inchi Code

1S/C11H16FNO/c1-8(14)7-13-9(2)10-4-3-5-11(12)6-10/h3-6,8-9,13-14H,7H2,1-2H3

Inchi Key

IHIXHAMAZLJOLN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library