Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C7H2Cl3NS/c8-3-1-5(10)11-6-4(9)2-12-7(3)6/h1-2H
Inchi Key
NICFXLUOWKFSRW-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 3,5,7-Trichlorothieno[3,2-b]pyridine
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
