Skip to Content

BDPH9BCDF75B

Bide Pharmatech

2-[(4-Methoxyphenyl)methyl]oxirane

Synonym(s):

2-(4-methoxybenzyl)oxirane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H12O2
CAS Number:
Molecular Weight:
164.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H12O2/c1-11-9-4-2-8(3-5-9)6-10-7-12-10/h2-5,10H,6-7H2,1H3

Inchi Key

CLOGCPPDCZMWGQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 2-[(4-Methoxyphenyl)methyl]oxirane



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library