Skip to Content
Merck

BDPH9BCDF91F

Bide Pharmatech

6-(Methylthio)pyridin-3-amine

Synonym(s):

6-(methylsulfanyl)-3-pyridinamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8N2S1
CAS Number:
Molecular Weight:
140.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8N2S/c1-9-6-3-2-5(7)4-8-6/h2-4H,7H2,1H3

Inchi Key

BEMNZDZGBAGRAT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to Pale-yellow to Brown Powder or Crystals



Still not finding the right product?

Explore all of our products under 6-(Methylthio)pyridin-3-amine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library