Skip to Content
Merck

BDPH9BCE0FA9

Bide Pharmatech

1-(4-Methoxyphenyl)ethylamine

Synonym(s):

1-(4-methoxyphenyl)ethylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H13O1N1
CAS Number:
Molecular Weight:
151.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H13NO/c1-7(10)8-3-5-9(11-2)6-4-8/h3-7H,10H2,1-2H3

Inchi Key

JTDGKQNNPKXKII-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 1-(4-Methoxyphenyl)ethylamine


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library