Skip to Content

BDPH9BCE13FF

Bide Pharmatech

tert-Butyl (3-cyano-4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-7-fluorobenzo[b]thiophen-2-yl)carbamate

Synonym(s):

tert-butyl (3-cyano-4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-7-fluorobenzo[b]thiophen-2-yl)carbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C19H22O4N2S1F1B1
CAS Number:
Molecular Weight:
404.27
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C19H22BFN2O4S/c1-18(2,3)27-17(24)23-16-11(8-22)14-12(6-7-13(21)15(14)28-16)20-25-9-19(4,5)10-26-20/h6-7H,9-10H2,1-5H3,(H,23,24)

Inchi Key

YGHCWKONJMTFIZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid






Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library