Skip to Content
Merck

Skip To

BDPH9C01E8FF

Bide Pharmatech

tert-Butyl ((1H-pyrazol-1-yl)((2,2,2-trifluoroacetyl)imino)methyl)carbamate

Synonym(s):

tert-butyl (Z)-((1H-pyrazol-1-yl)((2,2,2-trifluoroacetyl)imino)methyl)carbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C11H13O3N4F3
CAS Number:
Molecular Weight:
306.24
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid

Inchi Code

1S/C11H13F3N4O3/c1-10(2,3)21-9(20)17-8(18-6-4-5-15-18)16-7(19)11(12,13)14/h4-6H,1-3H3,(H,16,17,19,20)

Inchi Key

JLMUJVVPAITXSJ-UHFFFAOYSA-N

Shipping Temperature

Normal



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library