Skip to Content

BDPH9C01FEFB

Bide Pharmatech

6-Chloro-4-(trifluoromethyl)pyridazin-3(2H)-one

Synonym(s):

6-chloro-4-(trifluoromethyl)pyridazin-3-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H2O1N2Cl1F3
CAS Number:
Molecular Weight:
198.53
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H2ClF3N2O/c6-3-1-2(5(7,8)9)4(12)11-10-3/h1H,(H,11,12)

Inchi Key

QWYYINLSOLYYKF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library