Skip to Content
Merck

BDPH9C021A8D

Bide Pharmatech

(SP-4-1)-[[4,4',4'',4'''-(21H,23H-Porphine-5,10,15,20-tetrayl-κN21,κN22,κN23,κN24)tetrakis[benzoato]](6-)]manganate(3-)

Synonym(s):

monomanganese(III) mono((5Z,9Z,15Z,19Z)-5,10,15,20-tetrakis(4-carboxylatophenyl)-22H,24H-porphyrin-21,23-diide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9C021A8D is not currently sold in your country.


Inchi Code

1S/C48H30N4O8.Mn/c53-45(54)29-9-1-25(2-10-29)41-33-17-19-35(49-33)42(26-3-11-30(12-4-26)46(55)56)37-21-23-39(51-37)44(28-7-15-32(16-8-28)48(59)60)40-24-22-38(52-40)43(36-20-18-34(41)50-36)27-5-13-31(14-6-27)47(57)58;/h1-24,49,52H,(H,53,54)(H,55,56)(H,57,58)(H,59,60);/q-2;+3/p-4/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;

Inchi Key

OBLGLNOHDLWLDA-DAJBKUBHSA-J

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid






Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library