Skip to Content
Merck

BDPH9C021C28

Bide Pharmatech

1,2-Dimethyl-1H-imidazole-4-sulfonyl chloride

Synonym(s):

1,2-dimethyl-1H-imidazole-4-sulfonyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H7O2N2Cl1S1
Molecular Weight:
194.64
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H7ClN2O2S/c1-4-7-5(3-8(4)2)11(6,9)10/h3H,1-2H3

Inchi Key

JPAFTHVNSBWVQJ-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library