Skip to Content
Merck

BDPH9C021D71

Bide Pharmatech

(1S,3R)-3-Aminocyclohexanecarboxylic acid hydrochloride

Synonym(s):

(1S,3R)-3-aminocyclohexane-1-carboxylic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H14O2N1Cl1
CAS Number:
Molecular Weight:
179.65
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H13NO2.ClH/c8-6-3-1-2-5(4-6)7(9)10;/h5-6H,1-4,8H2,(H,9,10);1H/t5-,6+;/m0./s1

Inchi Key

BOUHXQPJYJANEO-RIHPBJNCSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to off-white solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library