Skip to Content
Merck

BDPH9C021E56

Bide Pharmatech

3-Phenylbut-2-enoic acid

Synonym(s):

(2E)-3-phenyl-2-butenoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O2
Molecular Weight:
162.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H10O2/c1-8(7-10(11)12)9-5-3-2-4-6-9/h2-7H,1H3,(H,11,12)/b8-7+

Inchi Key

PEXWJYDPDXUVSV-BQYQJAHWSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library