Skip to Content
Merck

BDPH9C024230

Bide Pharmatech

(2S,3S)-2-((S)-1-((2S,3S)-2-Amino-3-methylpentanoyl)pyrrolidine-2-carboxamido)-3-methylpentanoic acid

Synonym(s):

(2S,3S)-2-[({(2S)-1-[(2S,3S)-2-amino-3-methylpentanoyl]-2-pyrrolidinyl}carbonyl)amino]-3-methylpentanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H31O4N3
CAS Number:
Molecular Weight:
341.45
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H31N3O4/c1-5-10(3)13(18)16(22)20-9-7-8-12(20)15(21)19-14(17(23)24)11(4)6-2/h10-14H,5-9,18H2,1-4H3,(H,19,21)(H,23,24)/t10-,11-,12-,13-,14-/m0/s1

Inchi Key

JNTMAZFVYNDPLB-PEDHHIEDSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid






Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library