Inchi Code
1S/C6H2BrClFI/c7-4-1-3(8)2-5(10)6(4)9/h1-2H
Inchi Key
OAVPTVSBHAAKMS-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Country of Origin
CN
Physical Form
Solid
Storage temp.
Keep in dark place, sealed in dry, 2-8°C
Still not finding the right product?
Explore all of our products under 1-Bromo-5-chloro-2-fluoro-3-iodobenzene
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
