Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Ships in 14 days - from Aldrich Partner.
| € 35,00 |
5 g | Ships in 14 days - from Aldrich Partner.
| € 38,00 |
25 g | Ships in 14 days - from Aldrich Partner.
| € 58,00 |
100 g | Ships in 14 days - from Aldrich Partner.
| € 154,00 |
500 g | Ships in 14 days - from Aldrich Partner.
| € 612,00 |
About This Item
Vendor SKU: BD4732-1G
€ 35,00
Ships in 14 days - from Aldrich Partner.
Inchi Code
1S/C7H7BrClN/c1-4-6(10)3-2-5(8)7(4)9/h2-3H,10H2,1H3
Inchi Key
IAWSZYHMEPZGKK-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
96%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
solid
Still not finding the right product?
Explore all of our products under 4-Bromo-3-chloro-2-methylaniline
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
