Skip to Content
Merck

BL3H1F1CA02F

BLD Pharmatech

3,5-Dichloro-2-hydroxybenzaldehyde

Synonym(s):

3,5-dichloro-2-hydroxybenzaldehyde

Select a Size

Change View

About This Item

Linear Formula:
C7H4O2Cl2
CAS Number:
Molecular Weight:
191.01
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H4Cl2O2/c8-5-1-4(3-10)7(11)6(9)2-5/h1-3,11H

Inchi Key

FABVMBDCVAJXMB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: Very pale yellow - Pale yellow green - Yellow

Melting Point

95-97C



Still not finding the right product?

Explore all of our products under 3,5-Dichloro-2-hydroxybenzaldehyde


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library