Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 g | Ships in 14 days - from Aldrich Partner.
| € 48,00 |
100 g | Ships in 14 days - from Aldrich Partner.
| € 51,00 |
500 g | Ships in 14 days - from Aldrich Partner.
| € 54,00 |
1 kg | Ships in 14 days - from Aldrich Partner.
| € 100,00 |
About This Item
Vendor SKU: BD127403-25G
€ 48,00
Ships in 14 days - from Aldrich Partner.
Inchi Code
1S/C6H10O2/c7-6-4-2-1-3-5-8-6/h1-5H2
Inchi Key
PAPBSGBWRJIAAV-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under Oxepan-2-one
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
