Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
10 g | Ships in 14 days - from Aldrich Partner.
| € 48,00 |
25 g | Ships in 14 days - from Aldrich Partner.
| € 52,00 |
100 g | Ships in 14 days - from Aldrich Partner.
| € 57,00 |
500 g | Ships in 14 days - from Aldrich Partner.
| € 83,00 |
1 kg | Ships in 14 days - from Aldrich Partner.
| € 155,00 |
About This Item
Vendor SKU: BD69381-10G
€ 48,00
Ships in 14 days - from Aldrich Partner.
Purity
97%
Inchi Code
1S/C8H9NO2/c1-6(10)9-7-4-2-3-5-8(7)11/h2-5,11H,1H3,(H,9,10)
Inchi Key
ADVGKWPZRIDURE-UHFFFAOYSA-N
Shipping Temperature
Normal
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Form: Crystal - Powder / Colour: Very pale yellow - Pale reddish yellow - Brown
Melting Point
205-210C
Still not finding the right product?
Explore all of our products under N-(2-Hydroxyphenyl)acetamide
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
