Skip to Content
Merck

BL3H97A48EEE

BLD Pharmatech

1-Chloro-4-fluoro-2-iodobenzene

Synonym(s):

1-chloro-4-fluoro-2-iodobenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H3Cl1F1I1
CAS Number:
Molecular Weight:
256.45
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H3ClFI/c7-5-2-1-4(8)3-6(5)9/h1-3H

Inchi Key

RAHHKUWCBDUSPZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Liquid

Storage temp.

Keep in dark place, sealed in dry, 2-8C



Still not finding the right product?

Explore all of our products under 1-Chloro-4-fluoro-2-iodobenzene


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library