Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Ships in 14 days - from Aldrich Partner.
| € 277,00 |
About This Item
Vendor SKU: BD00806240-1G
€ 277,00
Ships in 14 days - from Aldrich Partner.
Inchi Code
1S/C4H7NO3.ClH/c6-4(7)3-1-8-2-5-3;/h3,5H,1-2H2,(H,6,7);1H
Inchi Key
QDWFSTLUZHSBAS-DFWYDOINSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under (S)-Oxazolidine-4-carboxylic acid hydrochloride
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
