Skip to Content
Merck

Skip To

BL3H9AE4F3BC

BLD Pharmatech

Vinyl dodecanoate

Synonym(s):

vinyl laurate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Ships in 14 days - from Aldrich Partner.
€ 20,00
100 g
Ships in 14 days - from Aldrich Partner.
€ 65,00

About This Item

Linear Formula:
C14H26O2
CAS Number:
Molecular Weight:
226.36
MDL number:

Vendor SKU: BD156476-25G

€ 20,00


Ships in 14 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H26O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h4H,2-3,5-13H2,1H3

Inchi Key

GLVVKKSPKXTQRB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library