Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C8H8N2O2
CAS Number:
Molecular Weight:
164.16
NACRES:
NA.24
UNSPSC Code:
41116107
EC Number:
200-835-2
eCl@ss:
25140904
Pricing and availability is not currently available.
grade
certified reference material
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1.2 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in acetonitrile
application(s)
environmental
format
single component solution
storage temp.
−20°C
SMILES string
COC1=C(C#N)C(=O)N(C)C=C1
InChI
1S/C8H8N2O2/c1-10-4-3-7(12-2)6(5-9)8(10)11/h3-4H,1-2H3
InChI key
PETSAYFQSGAEQY-UHFFFAOYSA-N
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
WGK
WGK 2
Flash Point (°F)
35.6 °F - closed cup
Flash Point (°C)
2 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

