Skip to Content
Merck

Skip To

NMIP1395

Heptachlor epoxide (exo)

NMI Australia

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C10H5Cl7O
CAS Number:
Molecular Weight:
389.32
UNSPSC Code:
41116107
EC Number:
213-831-0


agency

NMI Australia certified reference material

manufacturer/tradename

NMI Australia

format

neat

storage temp.

2-8°C

InChI

1S/C10H5Cl7O/c11-3-1-2(4-5(3)18-4)9(15)7(13)6(12)8(1,14)10(9,16)17/h1-5H

InChI key

ZXFXBSWRVIQKOD-UHFFFAOYSA-N



Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters