Skip to Content

System maintenance is scheduled for Sunday, October 11, 8:00 AM to 12:00 PM CST (Sunday, 11th October 2026, 03:00 PM to 07:00 PM CET). Quotes, price checks, and order status inquiries may be temporarily unavailable.

Merck

Skip To

NMIS002

5α-Androstan-3α,17β-diol-17β-glucuronide

NMI Australia

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C25H40O8
CAS Number:
Molecular Weight:
468.58
UNSPSC Code:
41116107
eCl@ss:
25140904


agency

NMI Australia certified reference material

manufacturer/tradename

NMI Australia

format

neat

storage temp.

−20°C



Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters