Skip to Content
Merck

COMH93D5FFCA

Combi-Blocks, Inc.

3,6-Dibromo-9-(2-naphthalenyl)-9h-carbazole

Synonym(s):

3,6-dibromo-9-(naphthalen-2-yl)-9H-carbazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C22H13N1Br2
Molecular Weight:
451.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C22H13Br2N/c23-16-6-9-21-19(12-16)20-13-17(24)7-10-22(20)25(21)18-8-5-14-3-1-2-4-15(14)11-18/h1-13H

Inchi Key

ZPMPPIIUVAZBBQ-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library