Skip to Content
Merck

Skip To

ENA321763243

Enamine

4-[(5-cyanopyridin-2-yl)oxy]benzoic acid

Synonym(s):

4-[(5-cyano-2-pyridinyl)oxy]benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Ships in 14 days - from Aldrich Partner.
€ 165,90
250 mg
Ships in 14 days - from Aldrich Partner.
€ 220,50
500 mg
Ships in 14 days - from Aldrich Partner.
€ 379,10
1 g
Ships in 14 days - from Aldrich Partner.
€ 503,90
2.5 g
Ships in 14 days - from Aldrich Partner.
€ 944,60
5 g
Ships in 14 days - from Aldrich Partner.
€ 1.374,90
10 g
Ships in 14 days - from Aldrich Partner.
€ 2.017,10

About This Item

Linear Formula:
C13H8O3N2
CAS Number:
Molecular Weight:
240.22
MDL number:

Vendor SKU: EN300-74809-100MG

€ 165,90


Ships in 14 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H8N2O3/c14-7-9-1-6-12(15-8-9)18-11-4-2-10(3-5-11)13(16)17/h1-6,8H,(H,16,17)

Inchi Key

IVUDWMJNAPABOO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95

Storage temp.

RT

Country of Origin

UA

Physical Form

powder



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library