Skip to Content
Merck

ENAH97A16F60

Enamine

{4-fluoro-2-oxabicyclo[2.1.1]hexan-1-yl}methanol

Sign In to View Organizational & Contract Pricing

Select a Size

Product ENAH97A16F60 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
132.13
MDL number:
Product ENAH97A16F60 is not currently sold in your country.

Iupac Name

(4-fluoro-2-oxabicyclo[2.1.1]hexan-1-yl)methanol

Inchi Code

1S/C6H9FO2/c7-5-1-6(2-5,3-8)9-4-5/h8H,1-4H2

InChI key

JEYRAORIAKZMTR-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1666290

MSDS

https://www.enaminestore.com/catalog/EN300-1666290


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library