Skip to Content
Merck

Skip To

MATH8735F2C5

Matrix Scientific

Pigment Violet 23

Synonym(s):

9,19-dichloro-5,15-diethyl-5,15-dihydrocarbazolo[3',4':5,6][1,4]oxazino[2,3-b]indolo[3,2-h]phenoxazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Ships in 14 days - from Aldrich Partner.
€ 104,20
5 g
Ships in 14 days - from Aldrich Partner.
€ 177,46
10 g
Ships in 14 days - from Aldrich Partner.
€ 243,75

About This Item

Linear Formula:
C34H22O2N4Cl2
Molecular Weight:
589.48
MDL number:

Vendor SKU: 099381-1G

€ 104,20


Ships in 14 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C34H22Cl2N4O2/c1-3-39-21-11-7-5-9-17(21)25-23(39)15-13-19-31(25)41-33-27(35)30-34(28(36)29(33)37-19)42-32-20(38-30)14-16-24-26(32)18-10-6-8-12-22(18)40(24)4-2/h5-16H,3-4H2,1-2H3

Inchi Key

QRHKNXDJFJHAMM-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library