Skip to Content
Merck

Skip To

1.16129

LiChrospher® 100 RP-8 (5 µm)

HPLC sorbent

Synonym(s):

TM=["LiChrospher"] C8 sorbent for HPLC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Check Cart for Availability
€ 2.130,00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52
Particle size:
5 μm
Matrix active group:
C8 (RP8), C8 bonding phase
Pore size:
100 Å pore size
Matrix:
fully porous particle

€ 2.130,00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

LiChrospher® 100 RP-8 (5 µm),

material

stainless steel column

agency

suitable for USP L7

product line

LiChrospher®

feature

endcapped: no

analyte functional class(es)

antibiotics, fat soluble vitamins, lipids, nutraceuticals and antioxidants, oils, pigments

analyte chemical class(es)

PCBs (aroclor identification), PFAS, aflatoxins, alcohols and aldehyde, alkaloids, amino acids, cannabinoids as tms derivatives, carotenoids, coloring compounds, essential oils, esters, fatty acids, flavonoids, ketones, metabolized steroids, nitrosamines, nucleosides, oligonucleotides, peptides, pesticides, phenols, phospholipids, phthalate esters, preservatives, proteins, steroids (cholesteryl esters), sulfonamides

extent of labeling

12.5% Carbon loading

parameter

60 °C max. temp.

technique(s)

HPLC: suitable, mass spectrometry (MS): suitable (Low - Medium)

surface area

350 m2/g

matrix

fully porous particle

matrix active group

C8 (RP8), C8 bonding phase

particle size

5 μm

pore size

100 Å pore size

operating pH range

2-7.5

bulk density

200‑400 kg/m3

separation technique

reversed phase

storage temp.

no temp limit

General description

LiChrospher® RP-8 is a reliable and versatile traditionally produced spherical silica gel carrier with C-8 reversed phase properties. It is well suited for the separation of neutral, acidic and weak basic compounds.

Application


  • Development and validation of a HPLC-UV method for the determination in didanosine tablets.: This study presents a validated HPLC-UV method optimized using LiChrospher® 100 RP-8 (5 µm) for analyzing didanosine in tablet formulations, offering a reliable tool for quality control in pharmaceutical environments (de Oliveira et al., 2005).

  • Determination of steroid hormones in oral contraceptives by high-performance liquid chromatography.: Utilizes LiChrospher® 100 RP-8 (5 µm) to effectively separate and quantify steroid hormones in oral contraceptive pills, demonstrating the column′s applicability in complex pharmaceutical analyses (Inês et al., 2002).

  • Determination of fluoroquinolone residues in animal tissues by liquid chromatography.: Explores the use of LiChrospher® 100 RP-8 (5 µm) for the sensitive detection of fluoroquinolone residues in animal tissues, highlighting its potential for food safety and veterinary medicine applications (Posyniak et al., 1999).

  • Rapid and simple determination of oxytetracycline in chicken products.: Demonstrates the column′s efficiency in rapidly quantifying oxytetracycline residues in chicken, ensuring compliance with food safety standards (Furusawa, 1999).

  • Crocheted ETFE-reactor for on-line post-column photoderivatization of diclofenac in high-performance liquid chromatography.: Describes an innovative application involving LiChrospher® 100 RP-8 (5 µm) in a post-column reactor setup to enhance the detection sensitivity of diclofenac, underlining the column′s versatility in advanced analytical techniques (Kuhlmann et al., 1997).


Analysis Note

Application test: passes test
Pressure: ≤ 45 bar
Theoretical Plates (N/m) (Toluene): ≥ 60000
Capacity factor (Toluene): ≥ 0.7
stability testing (performance): passes test
separation factor (PTH-Lysine/PHB-Propyl-Ester, selectivity test): 1.20 - 1.55
separation factor (Naphthalene/Digitoxigenin, selectivity test): 3.5 - 4.7
separation factor (PTH-Lysine/Digitoxigenin, selsctivity test): 1.70 - 2.25
Particle Size (d10): 4.8 - 5.5 µm
Particle Size (d50): 6.5 - 7.5 µm
Particle Size (d90): 9.0 - 10.5 µm

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters