Skip to Content
Merck

219364-M

Cathepsin B, Human Liver

Cathepsin B, Human Liver, CAS 9047-22-7, is a purified native cathepsin B from human liver, purified by affinity chromatography. Upregulated in many types of tumors.

Sign In to View Organizational & Contract Pricing.

Select a Size

5 U

€ 319,00

€ 319,00


Check Cart for Availability

Request a Bulk Order

About This Item

EC Number:
NACRES:
NA.77
UNSPSC Code:
12352202
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Cathepsin B, Human Liver, Cathepsin B, Human Liver, CAS 9047-22-7, is a purified native cathepsin B from human liver, purified by affinity chromatography. Upregulated in many types of tumors.

biological source

human liver

description

Merck USA index - 14, 1905

assay

>95% (SDS-PAGE and HPLC)

form

liquid

specific activity

≥50 units/mg protein

purified by

affinity chromatography

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

activity assay: suitable

pH range

3.5-6.0

suitability

suitable for molecular biology

application(s)

life science and biopharma

shipped in

wet ice

storage temp.

−70°C

Quality Level

Gene Information

human ... CTSB(1508)

Application

Cathepsin B, Human Liver has been used to assess its linker cleavage or proteolytic activity using an LC/MS MRM-based quantitation method.[1][2]

Biochem/physiol Actions

Cathepsin B facilitates non-specific, bulk proteolysis within the acidic conditions of the endosomal/lysosomal compartment. Here, they break down both intracellular and extracellular proteins. [3]Upregulation and secretion of cathepsin B have been shown to occur in many types of tumors and to correlate positively with their invasive and metastatic capabilities. Cathepsin B is shown to facilitate tumor invasion by dissolving extracellular barriers.

Disclaimer

Toxicity: Standard Handling (A)

General description

Research area: Cell Signaling

Cathepsin B, Human Liver, is a native cathepsin B from human liver, purified by affinity chromatography. Cathepsin B is a lysosomal protease with endo/exopeptidasecitation and a carboxydipeptidase activity. It is also known as cysteine cathepsin and belongs to the C1 family of papain-like enzymes. Cathepsin B like H, L, C, X, F, O and V is ubiquitously expressed in human tissues.[3]

Optimal pH: 3.5-6.0.

Other Notes

Kostoulas, G., et al. 1999. FEBS Lett. 455, 286. Berquim, I.M., and Sloane, B.F. 1996. Adv. Exp. Med. Biol.389, 281. Keppler, D., et al. 1994. Biochim. Biophys. Acta.1226, 117. Musil, D., et al. 1991. EMBO J.10, 2321. Sloane B.F. 1990. Semin. Cancer Biol.1, 137.
One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol Z-RR-βNA per min at 40°C, pH 6.0 in the presence of DTT.

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 50 mM sodium acetate buffer, 1 mM EDTA, pH 5.0.

Preparation Note

Following initial thaw, aliquot and freeze (-70°C).
Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service