About This Item
Skip To
Product Name
HDAC Inhibitor XXII, NCH51, The HDAC Inhibitor XXII, NCH51, also referenced under CAS 848354-66-5, controls the biological activity of HDAC. This small molecule/inhibitor is primarily used for Cell Structure applications.
InChI
1S/C20H26N2O2S2/c1-15(2)19(24)25-13-9-4-3-8-12-18(23)22-20-21-17(14-26-20)16-10-6-5-7-11-16/h5-7,10-11,14-15H,3-4,8-9,12-13H2,1-2H3,(H,21,22,23)
InChI key
MDYDGUOQFUQOGE-UHFFFAOYSA-N
assay
≥97% (HPLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
DMSO: 100 mg/mL
shipped in
ambient
storage temp.
−20°C
Quality Level
Disclaimer
Other Notes
Suzuki, T., et al. 2007. Bioorg. Med. Chem. Lett.17, 1558.
Suzuki, T., et al. 2005. J. Med. Chem.48, 1019.
Preparation Note
Legal Information
General description
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service