Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C19H17Cl3N4O3
CAS Number:
Molecular Weight:
455.72
UNSPSC Code:
12352200
PubChem Substance ID:
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated, protect from light
Pricing and availability is not currently available.
assay
≥97% (HPLC)
Quality Segment
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated, protect from light
color
beige
solubility
DMSO: 50 mg/mL
storage temp.
−20°C
SMILES string
COCCOC1=NN(C(=N1)C2=CC(=C(C=C2)Cl)Cl)C3=CC=C(C=C3)NC(=O)CCl
General description
A cell-permeable chloromethyl amide that inhibits MALT1 proteolytic activity (IC50 = 5.84 µM against MALT1 paracaspase domain dimer-catalyzed Ac-LRSR-AMC hydrolysis) in an irreversible manner via covalent interaction, displaying little or no activity against caspase-3, -8, or -9. Due to irreversible binding and effective intracellular accumulation (HBL-1 intracellular [MI-2]t = 2 h = 5 µM; Medium [MI-2]t = 0 = 0.2 µM), MI-2 is at least 250-times more effective than Z-VRPR-FMK in blocking constitutive NF-κB activity and c-Rel nuclear translocation in HBL-1 cultures (equal or better blockage by 200 nM MI-2 than by 50 µM Z-VRPR-FMK). Shown to selectively inhibit MALT1-dependent ABC-DLBCL proliferation, but not the MALT1-independent growths of ABC- (HLY-1 and U2932) or GCB-DLBCL (OCI-Ly1 and OCI-Ly7) cells, both in cultures in vitro (GI50 in 48 h = 200, 400, 400, and 500 nM, respectively, against HBL-1, OCI-Ly3, OCI-Ly10, and TMD8) and in mice in vivo (65%,, 61%, and 0% suppression, respectively, of HBL-1, TMD8, and OCI-Ly1 tumor expansion 24 h after 14 daily 25 mg/kg/day i.p. dosages) without any apparent toxicity to mice.
Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
MALT1
MALT1
Reversible: no
Packaging
Packaged under inert gas
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.
Other Notes
Fontan, L., et al. 2012. Cancer Cell22, 812.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.