Skip to Content
Merck

Skip To

521237

PDGFR Tyrosine Kinase Inhibitor VII

The PDGFR Tyrosine Kinase Inhibitor VII, also referenced under CAS 251356-45-3, controls the biological activity of PDGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PDGFR Tyrosine Kinase Inhibitor VII, (Z)-3-(2,4-Dimethyl-5-(2-oxo-1,2-dihydro-indol-3-ylidenemethyl-6-phenyl)-1H-pyrrol-3-yl)-propionic acid, VEGFR Tyrosine Kinase Inhibitor XXXI, VEGFR2 Kinase Inhibitor XXIX

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C24H22N2O3
CAS Number:
Molecular Weight:
386.44
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

dark orange

solubility

DMSO: 25 mg/mL, ethanol: insoluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C24H22N2O3/c1-14-18(10-11-23(27)28)15(2)25-21(14)13-20-19-9-8-17(12-22(19)26-24(20)29)16-6-4-3-5-7-16/h3-9,12-13,25H,10-11H2,1-2H3,(H,26,29)(H,27,28)/b20-13-

InChI key

APYYTEJNOZQZNA-MOSHPQCFSA-N

General description

A cell-permeable indolinone compound that acts as a potent, ATP site-targeting PDGFR inhibitor (IC50 = 10 nM against PDGFRβ), while affecting Flk-1/KDR/ VEGFR2, FGFR1, and EGFR only at much higher concentrations (IC50 = 0.14, 2.29, and >100 µM, respectively). Reported to selectively inhibit PDGFβ- over EGF-stimulated proliferation in EGFR-expressing NIH 3T3 cultures (IC50 = 0.11 and 21.9 µM, respectively) and exhibit much weaker potency against the proliferation of HUVEC cells upon FGF or VEGF stimulation (IC50 = 10 µM).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Sun, L., et al. 1999. J. Med. Chem.42, 5120.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters