Skip to Content
Merck

AB9602

Anti-Plexin-A1 Antibody

Chemicon®, from rabbit

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PLXNA1(5361)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4301102HPA007499AV49932
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

species reactivity

rat, human

species reactivity

mouse, human

species reactivity

human

species reactivity

mouse, rat, horse, dog, guinea pig, rabbit, human

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

UniProt accession no.

Q9UIW2

UniProt accession no.

Q9UIW2

UniProt accession no.

Q9UIW2

UniProt accession no.

O75051

Immunogen

Synthetic peptide from the semaphorin domain of human plexin-A1.

Application

Research Category
Neuroscience
Research Sub Category
Growth Cones & Axon Guidance
This Anti-Plexin-A1 Antibody is validated for use in WB for the detection of Plexin-A1.
Western blot: 1:1,000 on neonatal rat brain and COS-7 cells transfected with mouse Plexin-A1. The antibody reacts with the ~220 kDa protein. The suggested antibody diluent is PBS containing 5% non-fat milk and 0.04% Tween 20. Other bands may be present depending on the concentration of antibody and sample used.
Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Plexin-A1. The immunogen sequence has low homology with other plexin family members.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 50% glycerol, 1 mg/mL BSA and 0.05% sodium azide.

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Séverine Marcos et al.
Human molecular genetics, 26(11), 2006-2017 (2017-03-24)
The olfacto-genital syndrome (Kallmann syndrome) associates congenital hypogonadism due to gonadotropin-releasing hormone (GnRH) deficiency and anosmia. This is a genetically heterogeneous developmental disease with various modes of transmission, including oligogenic inheritance. Previous reports have involved defective cell signaling by semaphorin-3A

Questions

  1. I have a question regarding this antibody, I'm planning to run a western blot analysis on zebrafish lysates for PLXNA1, and I was wondering if this antibody would be compatible with zebrafish lysates

    1 answer
    1. This product is tested for suitability in rats and humans. Suitability in zebrafish has not been tested and would have to be determined by the end user.

      See the link below for publications that may be helpful:
      https://www.nature.com/articles/s41436-021-01196-9
      https://scholarworks.uvm.edu/cgi/viewcontent.cgi?article=1108&context=hcoltheses

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service