Select a Size
About This Item
Skip To
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
rat (based on 100% sequence homology), mouse (based on 100% sequence homology)
packaging
antibody small pack of 25 μg
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... MBD5(55777)
Related Categories
1 of 4
This Item | WH0055777M1 | ABE1321 | SAB1305225 |
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| biological source rabbit | biological source mouse | biological source rabbit | biological source rabbit |
| clone polyclonal | clone 4A12-1B6, monoclonal | clone polyclonal | clone polyclonal |
| species reactivity human | species reactivity human | species reactivity mouse, human | species reactivity mouse |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| technique(s) immunocytochemistry: suitable, western blot: suitable, immunohistochemistry: suitable (paraffin) | technique(s) immunofluorescence: suitable, indirect ELISA: suitable, western blot: 1-5 μg/mL | technique(s) immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), western blot: suitable | technique(s) western blot: 1:1000 |
General description
Immunogen
Application
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected MBD5 in HeLa, A431, and HEK293 cells.
Western Blotting Analysis: 2 µg/mL from a representative lot detected MBD5 in human liver tissue lysate.
Epigenetics & Nuclear Function
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected MBD5 in human prostate tissue sections.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


