Select a Size
About This Item
Skip To
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
feline (based on 100% sequence homology), bovine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), horse (based on 100% sequence homology), canine (based on 100% sequence homology), gorilla (based on 100% sequence homology), porcine (based on 100% sequence homology)
technique(s)
immunohistochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... FBXW7(55294)
1 of 4
This Item | F2055 | MABD127 | WH0222235M6 |
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form purified from hybridoma cell culture | antibody form purified antibody | antibody form purified immunoglobulin |
| Quality Level 100 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
| biological source rabbit | biological source mouse | biological source mouse | biological source mouse |
| technique(s) immunohistochemistry: suitable, western blot: suitable | technique(s) immunocytochemistry: 10-20 μg/mL using human A431 or HeLa cells, indirect ELISA: suitable, western blot: 5-10 μg/mL using human A431 cell extracts | technique(s) western blot: suitable | technique(s) indirect ELISA: suitable, western blot: 1-5 μg/mL |
| Gene Information human ... FBXW7(55294) | Gene Information human ... FBXW7(55294) | Gene Information human ... FBXO15(201456) | Gene Information human ... FBXL13(222235) |
| clone polyclonal | clone FB407, monoclonal | clone 3B11.1, monoclonal | clone 1F7, monoclonal |
General description
Immunogen
Application
Epigenetics & Nuclear Function
Nuclear Receptors
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 0.5 µg/mL of this antibody detected F-box/WD repeat-containing protein 7 in 10 µg of human breast cancer MDA-MB-231 and non-tumorigenic MCF-10A breast epithelial cell lysates.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
A major focus of breast cancer research is to understand the mechanisms responsible for disease progression and drug resistance. Toward that end, it has been found that approximately two thirds of all human breast carcinomas overexpress the Estrogen Receptor α (ERα) protein and it remains the primary pharmacological target for endocrine therapy1,2. The normal cellular function of ERα is as a transcription factor that mediates a wide variety of physiological processes, many of which are dependent upon phosphorylation of the receptor at specific amino acid residues3,4. Indeed, ERα is known to be phosphorylated at a multitude of different sites, yet how these all correlate to disease remains unclear5. Here, we interrogated multiple sites of ERα for phosphorylation status by screening an extensive panel of different breast cancer patient samples and other non-breast cancer tissue microarray (TMA) slide samples to determine their relevance to disease.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


