Merck
NA12/32-221
NovAseptic® Flow Through (1/2") Valve Body 180°
Valve Body - 316L and Electropolished with Butt end for welding
Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
UNSPSC Code:
23151800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Quality Level
parameter
-1-10 bar max. pressure (-14.5-145 psig) at 25 °C (atmospheric pressure)
-80-200 °C max. temp. (-112-392 °F)
General description
NovAseptic® Valves are engineered for optimal performance, reliability, durability and ease of maintenance. NovAseptic® Valves comply with cGMP Design Qualification
criteria for aseptic processing. This is why we choose to call it ‘‘Aseptic by Design.”
Application
NovAseptic® Valves are specifically designed for aseptic applications and comply with the most stringent cleanability and sterilizability requirements. The focus on aseptic design is a significant feature for all valves. Flow through valve is a valve designed for a wide range of applications in the aseptic process; including manifolds, take off and drainage valves.
Biochem/physiol Actions
Net volume (Valve cavity, diaphragm applied): 4mLNet weight (Approximate): Weight kg (lb): 0.1(0.22)Surface roughness: Internal surface Ra ≤ 0.38 μm (15 μin) External surface Ra ≤ 1.6 μm (63 μin)
Other Notes
All open ends on the valve body are protected with plastic covers on delivery and packaged in a closed box.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany